MilliporeSigma
  • Serotonin signaling suppresses the nutrition-responsive induction of an alternate male morph in horn polyphenic beetles.
Serotonin signaling suppresses the nutrition-responsive induction of an alternate male morph in horn polyphenic beetles.
Journal of experimental zoology. Part A, Ecological and integrative physiology (2020-09-23)
Daniel B Schwab, Keeley D Newsom, Armin P Moczek
ABSTRACT
Environment-responsive development contributes significantly to the phenotypic variation visible to selection and as such possesses the potential to shape evolutionary trajectories. However, evaluation of the contributions of developmental plasticity to evolutionary diversification necessitates an understanding of the developmental mechanisms underpinning plastic trait expression. We investigated the role of serotonin signaling in the regulation and evolution of horn polyphenism in the beetle genus Onthophagus. Specifically, we assessed the role of serotonin in development by determining whether manipulating serotonin biosynthesis during the larval stage alters body size, developmental rate, and the formation of relative adult trait size in traits characterized by minimal (genitalia), moderate (elytra), and pronounced (horns) nutrition-responsive development in O. taurus. Second, we assessed serotonin's role in evolution by replicating a subset of our approaches across four species reflecting ancestral as well as derived conditions. Lastly, we employed immunohistochemical approaches to begin assessing whether serotonin may be acting via the endocrine or nervous system. Our results show that pharmacological manipulation of serotonin signaling affects overall size, developmental rate, and the body size threshold separating alternate male morphs. Threshold body sizes were affected across species, regardless of the severity of horn polyphenism, and independent of the precise morphological location of horns. However, histological assessments suggest it is unlikely serotonin functions as a neurotransmitter and instead may rely on other mechanisms that remain to be identified. We discuss the most important implications of our results for our understanding of the evolution of and through plasticity in horned beetles and beyond.
Sigma-Aldrich
5-Hydroxy-L-tryptophan, powder
Sigma-Aldrich
α-Methyl-DL-tryptophan, crystalline
Supelco
Serotonin, analytical standard