MilliporeSigma
  • Rottlerin: Structure Modifications and KCNQ1/KCNE1 Ion Channel Activity.
Rottlerin: Structure Modifications and KCNQ1/KCNE1 Ion Channel Activity.
ChemMedChem (2020-04-28)
Marco Lübke, Julian A Schreiber, Thang Le Quoc, Florian Körber, Jasmin Müller, Sivatharushan Sivanathan, Veronika Matschke, Janina Schubert, Nathalie Strutz-Seebohm, Guiscard Seebohm, Jürgen Scherkenbeck
ABSTRACT
The slow delayed rectifier potassium current (IKs ) is formed by the KCNQ1 (Kv 7.1) channel, an ion channel of four α-subunits that modulates KCNE1 β-subunits. IKs is central to the repolarization of the cardiac action potential. Loss of function mutation reducing ventricular cardiac IKs cause the long-QT syndrome (LQTS), a disorder that predisposes patients to arrhythmia and sudden death. Current therapy for LQTS is inadequate. Rottlerin, a natural product of the kamala tree, activates IKs and has the potential to provide a new strategy for rational drug therapy. In this study, we show that simple modifications such as penta-acetylation or penta-methylation of rottlerin blunts activation activity. Total synthesis was used to prepare side-chain-modified derivatives that slowed down KCNQ1/KCNE1 channel deactivation to different degrees. A binding hypothesis of rottlerin is provided that opens the way to improved IKs activators as novel therapeutics for the treatment of LQTS.
Sigma-Aldrich
1,3-Propylene sulfite, 99%
Sigma-Aldrich
Ethylenediamine diacetate, 98%