MilliporeSigma
  • MiR-18a-5p contributes to enhanced proliferation and migration of PASMCs via targeting Notch2 in pulmonary arterial hypertension.
MiR-18a-5p contributes to enhanced proliferation and migration of PASMCs via targeting Notch2 in pulmonary arterial hypertension.
Life sciences (2020-06-26)
Renying Miao, Wanli Liu, Chaoran Qi, Yan Song, Yonggan Zhang, Yiqun Fu, Weiping Liu, Yuchang Lang, Yifei Zhang, Zhimin Zhang
ABSTRACT
This study is undertaken to investigate the role and molecular mechanisms of miR-18a-5p in regulating pulmonary arterial hypertension (PAH) pathogenesis. Gene expression and protein levels were determined by qRT-PCR and western blot, respectively; Cell counting kti-8 and Transwell migration assays were used to determine the biological functions of miR-18a-5p in pulmonary arterial smooth muscle cells (PASMCs); bioinformatics analysis, luciferase reporter assays were used to elucidate the mechanisms of miR-18a-5p. MiR-18a-5p was up-regulated in the clinical samples from PAH patients. PASMCs treated with hypoxia exhibited enhanced proliferative ability and upregulated miR-18a-5p expression. Knockdown of miR-18a-5p attenuated hypoxia-induced hyper-proliferation and enhanced migratory potential of PASMCs; while miR-18a-5p overexpression promoted PASMC proliferation and migration. Further mechanistic studies showed that Notch2 was a direct target of miR-18a-5p and was repressed by miR-18a-5p overexpression. The rescue studies indicated that Notch2 overexpression counteracted the enhanced proliferation and migration induced by miR-18a-5p mimics in PASMCs. Similarly, Notch2 overexpression also block the effects caused by hypoxia in PASMCs. Moreover, Notch2 expression was down-regulated in the PAH patients and was negatively correlated with miR-18a-5p expression. In vivo animal studies further revealed the up-regulation of miR-18a-5p and the down-regulation of Notch2 in the PAH rats. Collectively, this study identified the up-regulated miR-18a-5p in the PAH patients; our data suggest that miR-18a-5p contributes to the enhanced proliferation and migration of PASMCs via repressing Notch2 expression.
Sigma-Aldrich
MISSION® esiRNA, targeting human NOTCH2
Sigma-Aldrich
(Tyr[SO3H]27)Cholecystokinin fragment 26-33 Amide, ≥97% (HPLC), powder