MilliporeSigma
  • Gut Bacterial Species Distinctively Impact Host Purine Metabolites during Aging in Drosophila.
Gut Bacterial Species Distinctively Impact Host Purine Metabolites during Aging in Drosophila.
iScience (2020-09-12)
Toshitaka Yamauchi, Ayano Oi, Hina Kosakamoto, Yoriko Akuzawa-Tokita, Takumi Murakami, Hiroshi Mori, Masayuki Miura, Fumiaki Obata
ABSTRACT
Gut microbiota impacts the host metabolome and affects its health span. How bacterial species in the gut influence age-dependent metabolic alteration has not been elucidated. Here we show in Drosophila melanogaster that allantoin, an end product of purine metabolism, is increased during aging in a microbiota-dependent manner. Allantoin levels are low in young flies but are commonly elevated upon lifespan-shortening dietary manipulations such as high-purine, high-sugar, or high-yeast feeding. Removing Acetobacter persici in the Drosophila microbiome attenuated age-dependent allantoin increase. Mono-association with A. persici, but not with Lactobacillus plantarum, increased allantoin in aged flies. A. persici increased allantoin via activation of innate immune signaling IMD pathway in the renal tubules. On the other hand, analysis of bacteria-conditioned diets revealed that L. plantarum can decrease allantoin by reducing purines in the diet. These data together demonstrate species-specific regulations of host purine levels by the gut microbiome.
Sigma-Aldrich
Lysozyme from chicken egg white, BioUltra, lyophilized powder, ≥98% (SDS-PAGE), ≥40,000 units/mg protein
Sigma-Aldrich
Adenine hemisulfate salt, ≥99%