MilliporeSigma
  • PP1 promotes cyclin B destruction and the metaphase-anaphase transition by dephosphorylating CDC20.
PP1 promotes cyclin B destruction and the metaphase-anaphase transition by dephosphorylating CDC20.
Molecular biology of the cell (2020-08-07)
James Bancroft, James Holder, Zoë Geraghty, Tatiana Alfonso-Pérez, Daniel Murphy, Francis A Barr, Ulrike Gruneberg
ABSTRACT
Ubiquitin-dependent proteolysis of cyclin B and securin initiates sister chromatid segregation and anaphase. The anaphase-promoting complex/cyclosome and its coactivator CDC20 (APC/CCDC20) form the main ubiquitin E3 ligase for these two proteins. APC/CCDC20 is regulated by CDK1-cyclin B and counteracting PP1 and PP2A family phosphatases through modulation of both activating and inhibitory phosphorylation. Here, we report that PP1 promotes cyclin B destruction at the onset of anaphase by removing specific inhibitory phosphorylation in the N-terminus of CDC20. Depletion or chemical inhibition of PP1 stabilizes cyclin B and results in a pronounced delay at the metaphase-to-anaphase transition after chromosome alignment. This requirement for PP1 is lost in cells expressing CDK1 phosphorylation-defective CDC206A mutants. These CDC206A cells show a normal spindle checkpoint response and rapidly destroy cyclin B once all chromosomes have aligned and enter into anaphase in the absence of PP1 activity. PP1 therefore facilitates the metaphase-to-anaphase transition by promoting APC/CCDC20-dependent destruction of cyclin B in human cells.
Sigma-Aldrich
PP1, ≥98% (HPLC)
Sigma-Aldrich
Anti-Cdc20 Antibody, clone AR12, clone AR12, Chemicon®, from mouse
Sigma-Aldrich
Anti-α-Tubulin antibody, Mouse monoclonal, clone DM1A, purified from hybridoma cell culture
Sigma-Aldrich
Nocodazole, ≥99% (TLC), powder