MilliporeSigma
  • 2-n-Butyl-9-methyl-8-[1,2,3]triazol-2-yl-9H-purin-6-ylamine and analogues as A2A adenosine receptor antagonists. Design, synthesis, and pharmacological characterization.
2-n-Butyl-9-methyl-8-[1,2,3]triazol-2-yl-9H-purin-6-ylamine and analogues as A2A adenosine receptor antagonists. Design, synthesis, and pharmacological characterization.
Journal of medicinal chemistry (2005-10-28)
Patrizia Minetti, Maria Ornella Tinti, Paolo Carminati, Massimo Castorina, Maria Assunta Di Cesare, Stefano Di Serio, Grazia Gallo, Orlando Ghirardi, Fabrizio Giorgi, Luca Giorgi, Giovanni Piersanti, Francesca Bartoccini, Giorgio Tarzia
ABSTRACT
Two types of adenosine receptor ligands were designed, i.e., 9H-purine and 1H-imidazo[4,5-c]pyridines, to obtain selective A(2A) antagonists, and we report here their synthesis and binding affinities for the four adenosine receptor subtypes A(1), A(2A), A(2B) and A(3). The design was carried out on the basis of the molecular modeling of a number of potent adenosine receptor antagonists described in the literature. Three compounds (25b-d) showed an interesting affinity and selectivity for the A(2A) subtype. One of them, i.e., ST1535 (2-n-butyl-9-methyl-8-[1,2,3]triazol-2-yl-9H-purin-6-ylamine, 25b) (K(i) A(2A) = 6.6 nM, K(i) A(1)/A(2A) = 12; K(i) A(2B)/A(2A) = 58; K(i) A(3)/A(2A) > 160), was selected for in vivo study and shown to induce a dose-related increase in locomotor activity, suggestive of an A(2A) antagonist type of activity.
Sigma-Aldrich
Atenolol, ≥98% (TLC), powder
Sigma-Aldrich
S-(4-Nitrobenzyl)-6-thioinosine, ≥98%, solid
Supelco
L-Glutamic acid, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Sigma-Aldrich
L-Glutamic acid, FCC
Sigma-Aldrich
(±)-Baclofen, ≥98% (HPLC), solid
Sigma-Aldrich
L-Glutamic acid, from non-animal source, meets EP testing specifications, suitable for cell culture, 98.5-100.5%
Sigma-Aldrich
L-Glutamic acid, ReagentPlus®, ≥99% (HPLC)
Sigma-Aldrich
L-Glutamic acid, BioUltra, ≥99.5% (NT)