MilliporeSigma
  • Quantitative analyses of EGFR localization and trafficking dynamics in the follicular epithelium.
Quantitative analyses of EGFR localization and trafficking dynamics in the follicular epithelium.
Development (Cambridge, England) (2020-07-19)
Nicole T Revaitis, Matthew G Niepielko, Robert A Marmion, Eric A Klein, Benedetto Piccoli, Nir Yakoby
ABSTRACT
To bridge the gap between qualitative and quantitative analyses of the epidermal growth factor receptor (EGFR) in tissues, we generated an sfGFP-tagged EGF receptor (EGFR-sfGFP) in Drosophila The homozygous fly appears similar to wild type with EGFR expression and activation patterns that are consistent with previous reports in the ovary, early embryo, and imaginal discs. Using ELISA, we quantified an average of 1100, 6200 and 2500 receptors per follicle cell (FC) at stages 8/9, 10 and ≥11 of oogenesis, respectively. Interestingly, the spatial localization of the EGFR to the apical side of the FCs at early stages depended on the TGFα-like ligand Gurken. At later stages, EGFR localized to basolateral positions of the FCs. Finally, we followed the endosomal localization of EGFR in the FCs. The EGFR colocalized with the late endosome, but no significant colocalization of the receptor was found with the early endosome. The EGFR-sfGFP fly is an exciting new resource for studying cellular localization and regulation of EGFR in tissues.
Sigma-Aldrich
Anti-dEGF Receptor, Extracellular Domain antibody, Mouse monoclonal, clone C-273, purified from hybridoma cell culture