MilliporeSigma
  • Weeping candidate genes screened using comparative transcriptomic analysis of weeping and upright progeny in an F1 population of Prunus mume.
Weeping candidate genes screened using comparative transcriptomic analysis of weeping and upright progeny in an F1 population of Prunus mume.
Physiologia plantarum (2020-08-06)
Tian-Yu Mao, Huan-Huan Zhu, Yao-Yao Liu, Man-Zhu Bao, Jun-Wei Zhang, Qiang Fu, Cai-Feng Xiong, Jie Zhang
ABSTRACT
Weeping is a specific plant architecture with high ornamental value. Despite the considerable importance of the weeping habit to landscaping applications and knowledge of plant architecture biology, little is known regarding the underlying molecular mechanisms. In this study, growth and phytohormone content were analyzed among the progeny of different branch types in an F1 mapping population of Prunus mume with varying plant architecture. Bulked segregant RNA sequencing was conducted to compare differences among progeny at a transcriptional level. The weeping habit appears to be a complex process regulated by a series of metabolic pathways, with photosynthesis and flavonoid biosynthesis highly enriched in differentially expressed genes between weeping and upright progeny. Based on functional annotation and homologous analyses, we identified 30 candidate genes related to weeping that merit further analysis, including 10 genes related to IAA and GA3 biosynthesis, together with 6 genes related to secondary branch growth. The results of this study will facilitate further studies of the molecular mechanisms underlying the weeping habit in P. mume.
Sigma-Aldrich
Indole-3-acetic acid sodium salt, BioReagent, suitable for plant cell culture, ≥98%