MilliporeSigma
  • Long-Term Feeding of Dairy Goats with Broccoli By-Product and Artichoke Plant Silages: Milk Yield, Quality and Composition.
Long-Term Feeding of Dairy Goats with Broccoli By-Product and Artichoke Plant Silages: Milk Yield, Quality and Composition.
Animals : an open access journal from MDPI (2020-09-20)
Paula Monllor, Raquel Muelas, Amparo Roca, Alberto S Atzori, José Ramón Díaz, Esther Sendra, Gema Romero
ABSTRACT
The aim of this experiment was to study the effects of a 40% inclusion of broccoli by-product (BB) and artichoke plant (AP) silages in dairy goat diets on the milk yield, composition and animal health status during a full lactation. Feed consumption was lower in AP and BB animals due to their composition and higher moisture content, and BB animals showed a significant reduction in body weight. Milk from the BB treatment had the highest fat content, total solids and useful dry matter content (5.02, 13.9 and 8.39%, respectively). The Se level was slightly lower in AP and BB animals; however, the milk of these treatments was the lowest in Na and, in the case of BB animals, the richest in Ca (1267 mg/kg). Control and AP milk showed a similar fatty acid profile, although AP had a more beneficial aptitude for human health (lower ratio of n6/n3, 12.5). Plasma components, as metabolic parameters, were adequate for goats. It was concluded that a 40% inclusion of AP is an adequate solution to reduce the cost of feeding without harming the animals' health or performance and to improve the nutritional milk quality. It is necessary to lower the BB level of inclusion to increase feed consumption.