MilliporeSigma
  • Highly Parallel Quantification and Compartment Localization of Transcription Factors and Nuclear Proteins.
Highly Parallel Quantification and Compartment Localization of Transcription Factors and Nuclear Proteins.
Cell reports (2020-02-27)
Alexander J Federation, Vivek Nandakumar, Brian C Searle, Andrew Stergachis, Hao Wang, Lindsay K Pino, Gennifer Merrihew, Ying S Ting, Nicholas Howard, Tanya Kutyavin, Michael J MacCoss, John A Stamatoyannopoulos
ABSTRACT
Transcription factors and other chromatin-associated proteins are difficult to quantify comprehensively. Here, we combine facile nuclear sub-fractionation with data-independent acquisition mass spectrometry to achieve rapid, sensitive, and highly parallel quantification of the nuclear proteome in human cells. We apply this approach to quantify the response to acute degradation of BET bromodomains, revealing unexpected chromatin regulatory dynamics. The method is simple and enables system-level study of previously inaccessible chromatin and genome regulators.
Sigma-Aldrich
3-(1-Pyridinio)-1-propanesulfonate, ≥97.0% (N)
Sigma-Aldrich
Anti-CREBBP antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution