MilliporeSigma
  • Influence of dissolved organic matter on the accumulation, metabolite production and multi-biological effects of environmentally relevant fluoxetine in crucian carp (Carassius auratus).
Influence of dissolved organic matter on the accumulation, metabolite production and multi-biological effects of environmentally relevant fluoxetine in crucian carp (Carassius auratus).
Aquatic toxicology (Amsterdam, Netherlands) (2020-07-28)
Zhenhua Yan, Xiadong Zhang, Xuhui Bao, Xin Ling, Haohan Yang, Jianchao Liu, Guanghua Lu, Yong Ji
ABSTRACT
Fluoxetine is a widely prescribed antidepressant that has been frequently detected in aquatic environments and is associated with a series of neurological, behavioural and neuroendocrine disruptions in nontarget organisms. However, studies on its effects in fish under realistic environmental conditions are still limited. In this study, we determined the influences of an environmentally relevant concentration of fluoxetine (100 ng/L) on crucian carp (Carassius auratus) in the presence of dissolved organic matter (DOM). Endpoints that were assessed included accumulation of fluoxetine and metabolite formation as well as related biological responses involving neurotransmission and metabolic processes. Fluoxetine was significantly bioconcentrated in the fish brain and liver and largely transformed to the active metabolite norfluoxetine. Brain neurotransmission processes related to serotonin and choline and liver metabolic status were simultaneously altered. DOM added at 1 mg/L had no effect on the accumulation of fluoxetine or its metabolites in different tissues of the fish. However, at 10 mg/L DOM facilitated fluoxetine and norfluoxetine accumulation in the liver, brain, kidney, gill and bile tissues of the fish. The neuroendocrine-disrupting effects on fish caused by fluoxetine were also enhanced by the co-addition of DOM at 10 mg/L. Binding with fluoxetine and the inhibition of metabolic functions caused by DOM may be responsible for this increase in effects. These findings imply that at high concentrations DOM can increase the toxicity of environmentally relevant concentrations of fluoxetine to fish.
Sigma-Aldrich
Fluoxetine-d5 hydrochloride, 98 atom % D