MilliporeSigma
  • 6PGD Upregulation is Associated with Chemo- and Immuno-Resistance of Renal Cell Carcinoma via AMPK Signaling-Dependent NADPH-Mediated Metabolic Reprograming.
6PGD Upregulation is Associated with Chemo- and Immuno-Resistance of Renal Cell Carcinoma via AMPK Signaling-Dependent NADPH-Mediated Metabolic Reprograming.
The American journal of the medical sciences (2020-08-25)
Jun Cao, Xiaosong Sun, Xuejun Zhang, Dehong Chen
ABSTRACT
The essential role of 6-phosphogluconate dehydrogenase (6PGD), the enzyme catalyzing the oxidative pentose phosphate pathway, in tumor growth and metabolism has garnered attention in recent years. In this work, we are the first to demonstrate that aberrant activation of 6PGD is a feature in renal cell carcinoma (RCC) and is critically involved in renal carcinogenesis and chemo- and immuno-resistance. 6PGD expression and activity were systematically analyzed in normal and malignant renal cells and tissues. The roles of 6PGD and its downstream mechanism were investigated using gain-of-function and loss-of-function approaches. 6PGD expression and enzyme activity were increased in RCC cells and patients' samples. Activation of 6PGD via gain-of-function approach promoted growth of normal kidney but not RCC cells, and alleviated the efficacy of chemotherapeutic (e.g., 5-FU) and immunotherapeutic (e.g., IFN-α) agents. In contrast, 6PGD inhibition using siRNA knockdown and pharmacological inhibitor physcion augmented the inhibitory effects of 5-FU and IFN-α in RCC. Mechanistic studies demonstrated that 6PGD inhibition activated AMPK signaling, leading to ACC1 enzyme inhibition and reduction of lipid synthesis. In addition, 6PGD inhibition disrupted NADPH and NADH homeostasis in RCC cells as shown by the decreased level of NADPH and NADH, and suppressed SIRT-1 activity. AMPK inhibition by siRNA knockdown reversed the inhibitory effects of physcion, demonstrating that the effect of 6PGD inhibition is AMPK activation dependent. Our work provides preclinical evidence that 6PGD inhibition may represent a potential therapeutic strategy to augment the efficacy of RCC standard of care drugs.
Sigma-Aldrich
MISSION® esiRNA, targeting human PGD