MilliporeSigma
  • Analytical considerations for postmortem metabolomics using GC-high-resolution MS.
Analytical considerations for postmortem metabolomics using GC-high-resolution MS.
Analytical and bioanalytical chemistry (2019-11-24)
Lana Brockbals, Thomas Kraemer, Andrea E Steuer
ABSTRACT
Metabolomics studies that aim to qualitatively and quantitatively characterize the entirety of small endogenous biomolecules in an organism are widely conducted in the clinical setting. They also become more and more popular in the field of forensics (toxicology), e.g., to assist in postmortem investigations by objective postmortem interval estimation. However, other issues in postmortem toxicology, such as the phenomenon of (time-dependent) postmortem redistribution, have not yet been tackled by metabolomics studies. Hence, the aim of the current study was to develop an (un)targeted gas chromatography-high-resolution mass spectrometry-based method for endogenous metabolites as a tool for large-scale (un)targeted human postmortem metabolomics investigations (e.g., to objectively assess PMR) with thorough analytical evaluation of this method to ensure fitness-to-purpose in terms of reliability and robustness. This was achieved by using a targeted metabolite subset (n = 56) and a targeted processing workflow. Evaluation experiments have shown that using an artificial matrix (revised simulated body fluid (rSBF) + 5% bovine serum albumin (BSA)) for calibration purposes, all parameters lay within the scope of the method (sensitivity, selectivity, calibration model, accuracy, precision, processed sample stability, and extraction efficiency). When applying this method to large-scale studies, samples should be run in randomized order if analysis time is expected to exceed 18-24 h and potential biomarkers that are found with this method should be verified by a specialized, targeted method (e.g., by using standard addition in authentic matrix for quantification purposes). Overall, the current method can be successfully used for conduction of time-dependent postmortem metabolomics investigations. Graphical abstract.
Sigma-Aldrich
Lignoceric acid, ≥99% (GC)
Sigma-Aldrich
Pipecolinic acid, 98%
Sigma-Aldrich
DL-Cysteine, technical grade
Sigma-Aldrich
Inosine, ≥99% (HPLC)