MilliporeSigma
  • Activation of Paraventricular Melatonin Receptor 2 Mediates Melatonin-Conferred Cardioprotection Against Myocardial Ischemia/Reperfusion Injury.
Activation of Paraventricular Melatonin Receptor 2 Mediates Melatonin-Conferred Cardioprotection Against Myocardial Ischemia/Reperfusion Injury.
Journal of cardiovascular pharmacology (2020-05-21)
Chao Zhang, Jin-Bao Yang, Wei Quan, Ying-Da Feng, Jian-Yu Feng, Liu-Siyuan Cheng, Xiao-Qiang Li, Hui-Nan Zhang, Wen-Sheng Chen
ABSTRACT
Previous studies have shown that melatonin (Mel) can effectively ameliorate myocardial ischemia/reperfusion (MI/R) injury, but the mechanism is yet to be fully elucidated. Mel receptors are expressed in the paraventricular nucleus (PVN), which is also involved in regulating cardiac sympathetic nerve activity. The aim of this study was to examine whether Mel receptors in the PVN are involved in the protective effects of Mel against MI/R injury. The results of quantitative polymerase chain reaction, western blot, and immunofluorescence assays indicated that Mel receptor 2 (MT2) expression in the PVN was upregulated after MI/R. Intraperitoneal administration of Mel significantly improved post-MI/R cardiac function and reduced the infarct size, whereas shRNA silencing of MT2 in the PVN partially blocked this effect. Intraperitoneal administration of Mel reduced sympathetic nerve overexcitation caused by MI/R, whereas shRNA silencing of MT2 in the PVN partially diminished this effect. Furthermore, enzyme-linked immunosorbent assay and western blot results indicated that intraperitoneal administration of Mel lowered the levels of inflammatory cytokines in the PVN after MI/R injury, whereas the application of sh-MT2 in the PVN reduced this effect of Mel. Mel significantly reduced the levels of NF-κB after astrocyte oxygen and glucose deprivation/reoxygenation injury, and this effect was offset when MT2 was silenced. The above experimental results suggest that MT2 in the PVN partially mediated the protective effects of Mel against MI/R injury, and its underlying mechanisms may be related to postactivation amelioration of PVN inflammation and reduction of cardiac sympathetic nerve overexcitation.
Sigma-Aldrich
MISSION® esiRNA, targeting human MT2A