MilliporeSigma
  • RTH-149 Cell Line, a Useful Tool to Decipher Molecular Mechanisms Related to Fish Nutrition.
RTH-149 Cell Line, a Useful Tool to Decipher Molecular Mechanisms Related to Fish Nutrition.
Cells (2020-07-28)
Guillaume Morin, Karine Pinel, Karine Dias, Iban Seiliez, Florian Beaumatin
ABSTRACT
Nowadays, aquaculture provides more than 50% of fish consumed worldwide but faces new issues that challenge its sustainability. One of them relies on the replacement of fish meal (FM) in aquaculture feeds by other protein sources without deeply affecting the whole organism's homeostasis. Multiple strategies have already been tested using in vivo approaches, but they hardly managed to cope with the multifactorial problems related to the complexities of fish biology together with new feed formulations. In this context, rainbow trout (RT) is particularly concerned by these problems, since, as a carnivorous fish, dietary proteins provide the amino acids required to supply most of its energetic metabolism. Surprisingly, we noticed that in vitro approaches considering RT cell lines as models to study RT amino acid metabolism were never previously used. Therefore, we decided to investigate if, and how, three major pathways described, in other species, to be regulated by amino acid and to control cellular homeostasis were functional in a RT cell line called RTH-149-namely, the mechanistic Target Of Rapamycin (mTOR), autophagy and the general control nonderepressible 2 (GCN2) pathways. Our results not only demonstrated that these three pathways were functional in RTH-149 cells, but they also highlighted some RT specificities with respect to the time response, amino acid dependencies and the activation levels of their downstream targets. Altogether, this article demonstrated, for the first time, that RT cell lines could represent an interesting alternative of in vivo experimentations for the study of fish nutrition-related questions.
Product Description
Supelco
Halofuginone hydrobromide, VETRANAL®, analytical standard
Sigma-Aldrich
Chloroquine diphosphate salt, powder or crystals, 98.5-101.0% (EP)