MilliporeSigma
  • Pharmacological characterizations of recombinant human 5-HT1D alpha and 5-HT1D beta receptor subtypes coupled to adenylate cyclase inhibition in clonal cell lines: apparent differences in drug intrinsic efficacies between human 5-HT1D subtypes.
Pharmacological characterizations of recombinant human 5-HT1D alpha and 5-HT1D beta receptor subtypes coupled to adenylate cyclase inhibition in clonal cell lines: apparent differences in drug intrinsic efficacies between human 5-HT1D subtypes.
Naunyn-Schmiedeberg's archives of pharmacology (1996-08-01)
J M Zgombick, L E Schechter, N Adham, S A Kucharewicz, R L Weinshank, T A Branchek
ABSTRACT
Recombinant human 5-HT1D alpha and 5-HT1D beta receptor subtypes were stably expressed in NIH-3T3 fibroblasts (1D alpha cell line) and Y-1 adrenocortical tumor cells (1D beta cell line), respectively, for pharmacological evaluations of serotonergic compounds to inhibit forskolin-stimulated cAMP accumulation (FSCA). [3H]LSD saturation studies indicated that 5-HT1D receptor expression levels were slightly higher in the 1D beta cell line (Bmax = 1334 +/- 134 fmol/mg protein) than in the 1D alpha cell line (Bmax = 900 +/- 218 fmol/mg protein). 5-HT inhibited FSCA with similar potencies (EC50 approximately 2 nM) in both assay systems. The rank order of agonist potencies in both clonal cell lines matched their pharmacological profiles previously determined in binding studies: dihydroergotamine > or = 5-carboxamidotryptamine (5-CT) > LSD > or = 5-HT > sumatriptan > 1-naphthylpiperazine (1-NP) > yohimbine > 8-hydroxy-2-(di-n-propylamino)tetralin (8-OH DPAT) > 1-(2,5-dimethoxy-4-iodophenyl)-2-aminopropane (DOI), with Ki/EC50 ratios greater than unity. Methiothepin acted as a silent antagonist at both human 5-HT1D alpha and 5-HT1D beta receptors with apparent dissociation constants (Kb values) of 12 +/- 1 nM and 3 +/- 1 nM, respectively. Whereas GR 127,935, metergoline, DOI, and quipazine acted as full agonists in the 1D alpha cell line, these compounds behaved as partial agonists in the 1D beta cell line. To determine whether high levels of receptor reserve might mask partial agonist activity in the two second messenger assay systems, studies were performed using the irreversible receptor alkylating agent N-ethoxycarbonyl-2-ethoxy-1,2-dihydroquinoline (EEDQ). The relationships between receptor occupancy and inhibition of FSCA were determined for 5-HT, sumatriptan, and 1-NP in both clonal cell lines after partial receptor inactivation using Furchgott analysis. Hyperbolic relationships between receptor occupancy and second messenger response were determined for 5-HT in both transfected cell lines. Steep hyperbolic relationships were also found for sumatriptan and 1-NP in the 1D beta cell line whereas nearly linear relationships were observed for these two compounds in the 1D alpha cell line. Moreover, KA/EC50 ratios of these compounds were significantly larger in the 1D beta (10-32) as compared to the 1D alpha (0.9-2.5) cell line. These data are consistent with the hypothesis that the two heterologous expression systems contain a differential amount of receptor reserve. Despite the presence of an apparently larger-receptor reserve in the 1D beta cell line, GR 127,935, metergoline, DOI, and quipazine behaved as partial agonists. Although the potencies (EC50 values) of compounds matched their respective affinity constants (Ki values) for the closely-related 5-HT1D subtypes, differences in intrinsic activities were observed for a few compounds between the two 5-HT1D receptor expression systems. Since receptor reserve is dependent on the properties of both the assay system and drug, the observed variations in intrinsic activity, although influenced by the variable amounts of receptor reserve in the two transfected cell lines, reflect primarily system-independent differences in the intrinsic efficacy of the tested compounds at the two human 5-HT1D receptors. Higher intrinsic efficacies of compounds at the human 5-HT1D alpha receptor relative to the human 5-HT1D beta subtype may be responsible for the higher intrinsic activities observed in the 1D alpha cell line, even though receptor reserve is apparently lower in this system.
Sigma-Aldrich
Quipazine maleate salt