MilliporeSigma
  • β-Carbolines found in cigarette smoke elevate intracranial self-stimulation thresholds in rats.
β-Carbolines found in cigarette smoke elevate intracranial self-stimulation thresholds in rats.
Pharmacology, biochemistry, and behavior (2020-09-15)
Andrew C Harris, Peter Muelken, Mark G LeSage
ABSTRACT
Identifying novel constituents that contribute to tobacco addiction is essential for developing more effective treatments and informing FDA regulation of tobacco products. While preclinical data indicate that monoamine oxidase (MAO) inhibitors can have abuse liability or potentiate the addiction-related effects of nicotine, most of these studies have used clinical MAO inhibitors (e.g., tranylcypromine) that are not present in cigarette smoke. The primary goal of this study was to evaluate the abuse potential of the β-carbolines harmane, norharmane, and harmine - MAO inhibitors that are found in cigarette smoke - in an intracranial self-simulation (ICSS) model in rats. A secondary goal was to evaluate the ability of norharmane to influence nicotine's acute effects on ICSS. None of the β-carbolines lowered ICSS thresholds at any dose studied when administered alone, suggesting a lack of abuse liability. Rather, all three β-carbolines produced dose-dependent elevations in ICSS thresholds, indicating aversive/anhedonic effects. Harmane and harmine also elevated ICSS response latencies, suggesting a disruption of motor function, albeit with reduced potency compared to their ICSS threshold-elevating effects. Norharmane (2.5 mg/kg) modestly attenuated the effects of nicotine on ICSS thresholds. Our findings indicate that these β-carbolines produced only aversive/anhedonic effects in an ICSS model when administered alone, and that norharmane unexpectedly attenuated nicotines acute effects on ICSS. Future work evaluating the addiction-related effects of nicotine combined with these and other MAO inhibitors present in smoke may be useful for understanding the role of MAO inhibition in tobacco addiction and informing FDA tobacco regulation.
Sigma-Aldrich
Harmine hydrochloride, ≥98% (HPLC)