MilliporeSigma
  • Development and characterization of swine primary respiratory epithelial cells and their susceptibility to infection by four influenza virus types.
Development and characterization of swine primary respiratory epithelial cells and their susceptibility to infection by four influenza virus types.
Virology (2019-01-08)
Chithra C Sreenivasan, Milton Thomas, Linto Antony, Tristen Wormstadt, Michael B Hildreth, Dan Wang, Ben Hause, David H Francis, Feng Li, Radhey S Kaushik
ABSTRACT
Influenza viruses are a group of respiratory pathogens that have evolved into four different types: A, B, C, and D. A common feature is that all four types are capable of replicating and transmitting among pigs. Here, we describe the development of isogenous cell culture system from the swine respiratory tract to study influenza viruses. Phenotypic characterization of swine primary nasal turbinate, trachea and lung cells revealed high expression of cytokeratin and demonstrated tissue site dependent expression of tight junction proteins. Furthermore, lectin binding assay on these cells demonstrated higher levels of Sia2-6Gal than Sia2-3Gal receptors and supported the replication of influenza A, B, C, and D viruses to appreciable levels at both 33 and 37 °C, but replication competence was dependent on virus type or temperature used. Overall, these swine primary respiratory cells showed epithelial phenotype, which is suitable for studying the comparative biology and pathobiology of influenza viruses.
Sigma-Aldrich
Monoclonal Anti-Desmin antibody produced in mouse, clone DE-U-10, ascites fluid
Sigma-Aldrich
Monoclonal Anti-Actin, α-Smooth Muscle, clone 1A4, ascites fluid
Sigma-Aldrich
Monoclonal Anti-Vimentin antibody produced in mouse, clone VIM-13.2, ascites fluid
Sigma-Aldrich
IgG2a, Kappa from murine myeloma, clone UPC 10, purified immunoglobulin, buffered aqueous solution
Sigma-Aldrich
IgG1, Kappa from murine myeloma, clone MOPC 21, purified immunoglobulin, buffered aqueous solution
Sigma-Aldrich
IgM, Lambda from murine myeloma, clone MOPC 104E, purified immunoglobulin, buffered aqueous solution