MilliporeSigma
  • The flavonoid agathisflavone modulates the microglial neuroinflammatory response and enhances remyelination.
The flavonoid agathisflavone modulates the microglial neuroinflammatory response and enhances remyelination.
Pharmacological research (2020-06-14)
Monique Marylin Alves de Almeida, Francesca Pieropan, Larissa de Mattos Oliveira, Manoelito Coelho Dos Santos Junior, Jorge Mauricio David, Juceni Pereira David, Victor Diógenes A da Silva, Cleide Dos Santos Souza, Silvia Lima Costa, Arthur Morgan Butt
ABSTRACT
Myelin loss is the hallmark of the demyelinating disease multiple sclerosis (MS) and plays a significant role in multiple neurodegenerative diseases. A common factor in all neuropathologies is the central role of microglia, the intrinsic immune cells of the central nervous system (CNS). Microglia are activated in pathology and can have both pro- and anti-inflammatory functions. Here, we examined the effects of the flavonoid agathisflavone on microglia and remyelination in the cerebellar slice model following lysolecithin induced demyelination. Notably, agathisflavone enhances remyelination and alters microglial activation state, as determined by their morphology and cytokine profile. Furthermore, these effects of agathisflavone on remyelination and microglial activation were inhibited by blockade of estrogen receptor α. Thus, our results identify agathisflavone as a novel compound that may act via ER to regulate microglial activation and enhance remyelination and repair.
Sigma-Aldrich
Anti-Neurofilament 70 kDa Antibody, clone DA2, culture supernatant, clone DA2, Chemicon®
Sigma-Aldrich
L-α-Lysophosphatidylcholine from egg yolk, ≥99%, Type I, powder
Sigma-Aldrich
Anti-Myelin Basic Protein Antibody, a.a. 82-87, culture supernatant, clone 12, Chemicon®