MilliporeSigma
  • Thiol-norbornene reactions to improve natural rubber dispersion in cellulose nanofiber coatings.
Thiol-norbornene reactions to improve natural rubber dispersion in cellulose nanofiber coatings.
Carbohydrate polymers (2020-10-15)
Kendra Fein, Douglas W Bousfield, William M Gramlich
ABSTRACT
Cellulose nanofibrils (CNF) coatings are excellent grease barriers for biodegradable packaging. However, barrier properties are moisture sensitive, so hydrophobic components such as latexes or polymers are needed to impart moisture resistance, but incompatibility leads to poor dispersion. In this work, CNF modified with norbornenes was reacted with natural rubber (NR) latex in water to improve dispersion, coating formation, and coating moisture resistance by creating hybrid particles. Mixtures and reacted samples were coated through filtration onto paper. The hybrid particles improved NR retention and drainage rate as compared to the CNF/latex mixture. Hybrid particle coatings showed more uniform NR dispersion as compared to the mixtures, which formed NR and CNF layers. NR addition to coatings increased the water contact angle, reduced water absorption, and decreased the water vapor transmission rate. All coatings passed a 12-kit grease test before folding and the mixtures formed a crack-resistant CNF layer on the surface.
Sigma-Aldrich
cis-5-Norbornene-endo-2,3-dicarboxylic anhydride, 99%
Sigma-Aldrich
2,2′-(Ethylenedioxy)diethanethiol, 95%