MilliporeSigma
  • Identifying Cranberry Juice Consumers with Predictive OPLS-DA Models of Plasma Metabolome and Validation of Cranberry Juice Intake Biomarkers in a Double-Blinded, Randomized, Placebo-Controlled, Cross-Over Study.
Identifying Cranberry Juice Consumers with Predictive OPLS-DA Models of Plasma Metabolome and Validation of Cranberry Juice Intake Biomarkers in a Double-Blinded, Randomized, Placebo-Controlled, Cross-Over Study.
Molecular nutrition & food research (2020-04-14)
Shaomin Zhao, Haiyan Liu, Zhihua Su, Christina Khoo, Liwei Gu
ABSTRACT
Methods to verify cranberry juice consumption are lacking. Predictive multivariate models built upon validated biomarkers may help to verify human consumption of a food using a nutrimetabolomics approach. A 21-day double-blinded, randomized, placebo-controlled, cross-over study was conducted among healthy young women aged 1829. Plasma was collected at baseline and after 3-day and 21-day consumption of cranberry or placebo juice. Plasma metabolome was analyzed using UHPLC coupled with high resolution mass spectrometry. 18 discriminant metabolites in positive mode and 18 discriminant metabolites in negative mode from a previous 3-day open-label study were re-discovered in the present blinded study. Predictive orthogonal partial least squares discriminant analysis (OPLS-DA) models were able to identify cranberry juice consumers over a placebo juice group with 96.9% correction rates after 3-day consumption in both positive and negative mode. This present study revealed 84 and 109 additional discriminant metabolites in positive and negative mode, respectively. Twelve of them were tentatively identified. Cranberry juice consumers were classified with high correction rates using predictive OPLS-DA models built upon validated plasma biomarkers. Additional biomarkers were tentatively identified. These OPLS-DA models and biomarkers provided an objective approach to verify participant compliance in future clinical trials.
Sigma-Aldrich
L-Leucine-2,3,3,4,5,5,5,5′,5′,5′-d10, 98 atom % D