MilliporeSigma
  • Tumor promoting properties of a cigarette smoke prevalent polycyclic aromatic hydrocarbon as indicated by the inhibition of gap junctional intercellular communication via phosphatidylcholine-specific phospholipase C.
Tumor promoting properties of a cigarette smoke prevalent polycyclic aromatic hydrocarbon as indicated by the inhibition of gap junctional intercellular communication via phosphatidylcholine-specific phospholipase C.
Cancer science (2008-04-02)
Brad L Upham, Ludek Bláha, Pavel Babica, Joon-Suk Park, Iva Sovadinova, Charles Pudrith, Alisa M Rummel, Liliane M Weis, Kimie Sai, Patti K Tithof, Miodrag Guzvić, Jan Vondrácek, Miroslav Machala, James E Trosko
ABSTRACT
Inhibition of gap junctional intercellular communication (GJIC) and the activation of intracellular mitogenic pathways are common hallmarks of epithelial derived cancer cells. We previously determined that the 1-methyl and not the 2-methyl isomer of anthracene, which are prominent cigarette smoke components, activated extracellular receptor kinase, and inhibited GJIC in WB-F344 rat liver epithelial cells. Using these same cells, we show that an immediate upstream response to 1-methylanthracene was a rapid (<1 min) release of arachidonic acid. Inhibition of phosphatidylcholine-specific phospholipase C prevented the inhibition of GJIC by 1-methylanthracene. In contrast, inhibition of phosphatidylinositol specific phospholipase C, phospholipase A(2), diacylglycerol lipase, phospholipase D, protein kinase C, and tyrosine protein kinases had no effect on 1-methylanthracene-induced inhibition of GJIC. Inhibition of protein kinase A also prevented inhibition of GJIC by 1-methylanthracene. Direct measurement of phosphatidylcholine-specific phospholipase C and sphingomyelinase indicated that only phosphatidylcholine-specific phospholipase C was activated in response to 1-methylanthracene, while 2-methylanthracene had no effect. 1-methylanthracene also activated p38-mitogen activated protein kinase; however, like extracellular kinase, its activation was not involved in 1-methylanthracene-induced regulation of GJIC, and this activation was independent of phosphatidylcholine-specific phospholipase C. Although mitogen activated protein kinases were activated, Western blot analyzes indicated no change in connexin43 phosphorylation status. Our results indicate that phosphatidylcholine-specific phospholipase C is an important enzyme in the induction of a tumorigenic phenotype, namely the inhibition of GJIC; whereas mitogen activated protein kinases triggered in response to 1-methylanthracene, were not involved in the deregulation of GJIC.
Sigma-Aldrich
Genistein, from Glycine max (soybean), ~98% (HPLC)
Sigma-Aldrich
Genistein, synthetic, ≥98% (HPLC), powder
Sigma-Aldrich
2-Methylanthracene, 97%
Supelco
Genistein, analytical standard