MilliporeSigma
  • In-situ polymerized polypyrrole nanoparticles immobilized poly(ε-caprolactone) electrospun conductive scaffolds for bone tissue engineering.
In-situ polymerized polypyrrole nanoparticles immobilized poly(ε-caprolactone) electrospun conductive scaffolds for bone tissue engineering.
Materials science & engineering. C, Materials for biological applications (2020-10-01)
Bikendra Maharjan, Vignesh Krishnamoorthi Kaliannagounder, Se Rim Jang, Ganesh Prasad Awasthi, Deval Prasad Bhattarai, Ghizlane Choukrani, Chan Hee Park, Cheol Sang Kim
ABSTRACT
Despite intensive attempts to fabricate polypyrrole nanoparticles (PPy-NPs) incorporated nanofibrous scaffolds, a low-cost facile strategy is still demanded. Herein, we developed a novel strategy- in-situ polymerization of PPy-NPs and immobilized them into the PCL polymeric matrix in a single step. For the in-situ polymerization of PPy-NPs, ferric chloride hexahydrate (FeCl3.6H2O) was introduced as an oxidant into the blended solution of PCL and pyrrole monomers. Due to the chemical oxidative polymerization process, the clear solution changed into a black PCL/PPy solution. After electrospinning the solution, PCL/PPy composite nanofibers were fabricated. The immobilization of PPy-NPs into PCL matrix was clearly revealed by Bio-TEM images. The Field emission scanning electron microscopy (FESEM) results exhibited that the PCL/PPy scaffolds showed significantly decreased fiber diameter. The atomic force microscopy (AFM) study showed increased surface roughness in the PCL/PPy scaffolds. The mechanical strength test of PCL/PPy scaffolds showed improved Young's Modulus (YM = 2 to 4-folds) and tensile strength (TS = 3 to 4-folds). As well as the YM and TS were gradually increased with increased concentration of PPy-NPs in composite scaffolds. The conductivity measurement conducted on polymeric solution and electrospun scaffolds showed an increasing trend of conductive property in the PCL/PPy solution and scaffolds too. The surface wettability test exhibited decreased water contact angle measurement from 126° for pure PCL to 93° for the PCL/PPy-200 composite scaffold. The biomineralization test conducted by simulated body fluid (SBF) incubation showed enhanced calcium-phosphate crystal deposition on the PCL/PPy scaffolds. The CCK-8 assay and confocal laser scanning microscopic (CLSM) imaging conducted without and with electrical stimulation (ES) displayed enhanced cell adhesion, growth, and proliferation of MC3T3-E1 cells on the PCL/PPy conductive scaffolds. Furthermore, ALP and ARS staining assays showed significant enhancement of the calcium-phosphate deposition on the PCL/PPy scaffolds after ES treatment. Hence, the current study provides a novel strategy for the fabrication of PCL/PPy conductive scaffolds with enhanced bioactivity, biocompatibility, and osteogenic differentiation under electrical stimulation confirmed its promising application towards bone tissue engineering.
Product Description
SAFC
Hanks′ Balanced Salts, Modified, without calcium chloride, magnesium sulfate and sodium bicarbonate, powder, suitable for cell culture