MilliporeSigma
  • Common insecticide disrupts aquatic communities: A mesocosm-to-field ecological risk assessment of fipronil and its degradates in U.S. streams.
Common insecticide disrupts aquatic communities: A mesocosm-to-field ecological risk assessment of fipronil and its degradates in U.S. streams.
Science advances (2020-10-25)
Janet L Miller, Travis S Schmidt, Peter C Van Metre, Barbara J Mahler, Mark W Sandstrom, Lisa H Nowell, Daren M Carlisle, Patrick W Moran
ABSTRACT
Insecticides in streams are increasingly a global concern, yet information on safe concentrations for aquatic ecosystems is sparse. In a 30-day mesocosm experiment exposing native benthic aquatic invertebrates to the common insecticide fipronil and four degradates, fipronil compounds caused altered emergence and trophic cascades. Effect concentrations eliciting a 50% response (EC50) were developed for fipronil and its sulfide, sulfone, and desulfinyl degradates; taxa were insensitive to fipronil amide. Hazard concentrations for 5% of affected species derived from up to 15 mesocosm EC50 values were used to convert fipronil compound concentrations in field samples to the sum of toxic units (∑TUFipronils). Mean ∑TUFipronils exceeded 1 (indicating toxicity) in 16% of streams sampled from five regional studies. The Species at Risk invertebrate metric was negatively associated with ∑TUFipronils in four of five regions sampled. This ecological risk assessment indicates that low concentrations of fipronil compounds degrade stream communities in multiple regions of the United States.
Product Description
Supelco
Fipronil sulfide, PESTANAL®, analytical standard