MilliporeSigma
  • Curcumin-loaded poly(ϵ-caprolactone) lipid-core nanocapsules: Evaluation of fetal and maternal toxicity.
Curcumin-loaded poly(ϵ-caprolactone) lipid-core nanocapsules: Evaluation of fetal and maternal toxicity.
Food and chemical toxicology : an international journal published for the British Industrial Biological Research Association (2020-08-02)
Renata Giacomeli, Flavia Elizabete Guerra Teixeira, Felipe Barbosa Carvalho, Camila Oliveira Pacheco, Alcides José Martins Parisotto, Ana Claudia Funguetto Ribeiro, Marcelo Gomes de Gomes, Sandra Elisa Haas
ABSTRACT
This study was designed to examine fetal and maternal toxicity of curcumin (CURC) loaded lipid-core nanocapsules (LNC) prepared with poly(ϵ-caprolactone) as a polymer, administered during the organogenesis period. Free CURC and CURC loaded-LNC (C-LNC) (2 mg/kg), blank LNC (B-LNC) and saline (CONTROL) were administered per oral route from the 7° to 13° gestational day (GD). Dams were evaluated daily for body weight gain, clinical signs, water and food intake. On 20° GD, dams were euthanized, organs were weighed and blood was collected for biochemical determinations. Fetal biometrics and external morphological anomalies were assessed. Also, were performed histopathological analysis of placenta and measurement of cytokines levels in placental and fetal liver tissues. All groups did not cause changes in dams during the pregnancy. Furthermore, treatments did not cause external morphological changes and delayed fetal development. Still, for histopathological analysis of placental tissue, treatments did not cause alterations in evaluated parameters. For cytokines levels, CURC and C-LNC caused a decrease in placental levels of TNF-α. Therefore, we have demonstrated that C-LNC did not cause toxicological effects (mother and fetus), in the same manner as pattern bioactive compound, proving to be a promising nutraceutical delivery system for maternal supplementation with CURC.
Sigma-Aldrich
Rat IL-10 ELISA Kit, for cell and tissue lysates
Sigma-Aldrich
Rat Interleukin-6 ELISA Kit, for for cell lysate and tissue lysate
Sigma-Aldrich
Sorbitan monostearate, meets FCC analytical specifications
Sigma-Aldrich
Rat Tumor Necrosis Factor α ELISA Kit, for cell and tissue lysates
Sigma-Aldrich
Rat IL-1 β ELISA Kit, for cell and tissue lysates