MilliporeSigma
  • Room-temperature formation of CdS magic-size clusters in aqueous solutions assisted by primary amines.
Room-temperature formation of CdS magic-size clusters in aqueous solutions assisted by primary amines.
Nature communications (2020-08-23)
Wushuang Wan, Meng Zhang, Min Zhao, Nelson Rowell, Chunchun Zhang, Shanling Wang, Theo Kreouzis, Hongsong Fan, Wen Huang, Kui Yu
ABSTRACT
Aqueous-phase approaches to semiconductor CdS magic-size clusters (MSCs) and the formation pathway have remained relatively unexplored. Here, we report the demonstration of an aqueous-phase, room-temperature approach to CdS MSCs, together with an exploration of their evolution pathway. The resulting CdS MSCs display a sharp optical absorption peak at about 360 nm and are labeled MSC-360. With CdCl2 and thiourea as the respective Cd and S sources, and 3-mercarpotopropionic acid as the ligand, CdS MSC-360 develops in a mixture of a primary amine and water. We argue that the primary amine facilitates room-temperature decomposition of thiourea when CdCl2 is present, and the formation pathway of MSCs is similar to that in organic-phase approaches. Our findings show there is a viable avenue to room-temperature aqueous-phase formation of CdS MSCs. Providing explanations of the procedure developed including the formation of large aggregates, the present study represents an important advance towards a mechanistic understanding of nanocrystal synthesis.
Sigma-Aldrich
4,4,4-Trifluoro-1-phenyl-1,3-butanedione, 99%
Sigma-Aldrich
Pyrene, sublimed grade, 99%