MilliporeSigma
  • Autonomous mesoscale positioning emerging from myelin filament self-organization and Marangoni flows.
Autonomous mesoscale positioning emerging from myelin filament self-organization and Marangoni flows.
Nature communications (2020-09-25)
Arno van der Weijden, Mitch Winkens, Sandra M C Schoenmakers, Wilhelm T S Huck, Peter A Korevaar
ABSTRACT
Out-of-equilibrium molecular systems hold great promise as dynamic, reconfigurable matter that executes complex tasks autonomously. However, translating molecular scale dynamics into spatiotemporally controlled phenomena emerging at mesoscopic scale remains a challenge-especially if one aims at a design where the system itself maintains gradients that are required to establish spatial differentiation. Here, we demonstrate how surface tension gradients, facilitated by a linear amphiphile molecule, generate Marangoni flows that coordinate the positioning of amphiphile source and drain droplets floating at air-water interfaces. Importantly, at the same time, this amphiphile leads, via buckling instabilities in lamellar systems of said amphiphile, to the assembly of millimeter long filaments that grow from the source droplets and get absorbed at the drain droplets. Thereby, the Marangoni flows and filament organization together sustain the autonomous positioning of interconnected droplet-filament networks at the mesoscale. Our concepts provide potential for the development of non-equilibrium matter with spatiotemporal programmability.
Sigma-Aldrich
Sodium oleate, ≥95% (capillary GC)