MilliporeSigma
Washable Colorimetric Nanofiber Nonwoven for Ammonia Gas Detection.
Polymers (2020-07-28)
Hyun Ju Oh, Byeong Jin Yeang, Young Ki Park, Hyun Jung Choi, Jong H Kim, Young Sik Kang, Younghwan Bae, Jung Yeon Kim, Seung Ju Lim, Woosung Lee, Wan-Gyu Hahm
ABSTRACT
The colorimetric sensor is a facile, cost-effective, and non-power-operated green energy material for gas detection. In this study, the colorimetric sensing property of a meta-aramid/dye 3 nanofiber sensor for ammonia (NH3) gas detection was investigated. This colorimetric sensor was prepared using various dye 3 concentrations via electrospinning. Morphological, thermal, structural, and mechanical analyses of the sensor were carried out by field-emission scanning electron microscopy, thermogravimetric analysis, Fourier-transform infrared spectroscopy, and a universal testing machine, respectively. A homemade computer color matching machine connected with a gas flow device characterized the response of the meta-aramid/dye 3 nanofiber colorimetric sensor to various exposure levels of NH3 gas. From the results, we confirmed that this colorimetric green energy sensor could detect ammonia gas in the concentration of 1-10 ppm with a sensing response time of 10 s at room temperature. After washing with laundry detergent for 30 min, the colorimetric sensors still exhibited sensing property and reversibility.
Sigma-Aldrich
Isophthaloyl chloride, ≥99%