MilliporeSigma
  • An optimized acetylcholine sensor for monitoring in vivo cholinergic activity.
An optimized acetylcholine sensor for monitoring in vivo cholinergic activity.
Nature methods (2020-09-30)
Miao Jing, Yuexuan Li, Jianzhi Zeng, Pengcheng Huang, Miguel Skirzewski, Ornela Kljakic, Wanling Peng, Tongrui Qian, Ke Tan, Jing Zou, Simon Trinh, Runlong Wu, Shichen Zhang, Sunlei Pan, Samuel A Hires, Min Xu, Haohong Li, Lisa M Saksida, Vania F Prado, Timothy J Bussey, Marco A M Prado, Liangyi Chen, Heping Cheng, Yulong Li
ABSTRACT
The ability to directly measure acetylcholine (ACh) release is an essential step toward understanding its physiological function. Here we optimized the GRABACh (GPCR-activation-based ACh) sensor to achieve substantially improved sensitivity in ACh detection, as well as reduced downstream coupling to intracellular pathways. The improved version of the ACh sensor retains the subsecond response kinetics, physiologically relevant affinity and precise molecular specificity for ACh of its predecessor. Using this sensor, we revealed compartmental ACh signals in the olfactory center of transgenic flies in response to external stimuli including odor and body shock. Using fiber photometry recording and two-photon imaging, our ACh sensor also enabled sensitive detection of single-trial ACh dynamics in multiple brain regions in mice performing a variety of behaviors.
Sigma-Aldrich
Isopentyl acetate, anhydrous, ≥99%