MilliporeSigma
  • Enhanced protocol for quantitative N-linked glycomics analysis using Individuality Normalization when Labeling with Isotopic Glycan Hydrazide Tags (INLIGHT)™.
Enhanced protocol for quantitative N-linked glycomics analysis using Individuality Normalization when Labeling with Isotopic Glycan Hydrazide Tags (INLIGHT)™.
Analytical and bioanalytical chemistry (2020-08-28)
Jaclyn Gowen Kalmar, Karen E Butler, Erin S Baker, David C Muddiman
ABSTRACT
The analysis of N-linked glycans using liquid chromatography and mass spectrometry (LC-MS) presents significant challenges, particularly owing to their hydrophilic nature. To address these difficulties, a variety of derivatization methods have been developed to facilitate improved ionization and detection sensitivity. One such method, the Individuality Normalization when Labeling with Isotopic Glycan Hydrazide Tags (INLIGHT)™ strategy for labeling glycans, has previously been utilized in the analysis of N- and O-linked glycans in biological samples. To assess the maximum sensitivity and separability of the INLIGHT™ preparation and analysis pipeline, several critical steps were investigated. First, recombinant and nonrecombinant sources of PNGase F were compared to assess variations in the released glycans. Second, modifications in the INLIGHT™ derivatization step were evaluated including temperature optimization, solvent composition changes, reaction condition length and tag concentration. Optimization of the modified method resulted in 20-100 times greater peak areas for the detected N-linked glycans in fetuin and horseradish peroxidase compared with the standard method. Furthermore, the identification of low-abundance glycans, such as (Fuc)1(Gal)2(GlcNAc)4(Man)3(NeuAc)1 and (Gal)3(GlcNAc)5(Man)3(NeuAc)3, was possible. Finally, the optimal LC setup for the INLIGHT™ derivatized N-linked glycan analyses was found to be a C18 reverse-phase (RP) column with mobile phases typical of RPLC.
Sigma-Aldrich
N-[Tris(hydroxymethyl)methyl]acrylamide, contains ≤7% KCl, 93%
Carbon, tube, outside diameter 3.18 mm, length 150 mm, wall thickness 0.8 mm