MilliporeSigma
  • Sugar causes obesity and metabolic syndrome in mice independently of sweet taste.
Sugar causes obesity and metabolic syndrome in mice independently of sweet taste.
American journal of physiology. Endocrinology and metabolism (2020-06-24)
Ana Andres-Hernando, Masanari Kuwabara, David J Orlicky, Aurelie Vandenbeuch, Christina Cicerchi, Sue C Kinnamon, Thomas E Finger, Richard J Johnson, Miguel A Lanaspa
ABSTRACT
Intake of sugars, especially the fructose component, is strongly associated with the development of obesity and metabolic syndrome, but the relative role of taste versus metabolism in driving preference, intake, and metabolic outcome is not fully understood. We aimed to evaluate the preference for sweet substances and the tendency to develop metabolic syndrome in response to these sugars in mice lacking functional taste signaling [P2X2 (P2X purinoreceptor 2)/P2X3 (P2X purinoreceptor 3) double knockout mice (DKO)] and mice unable to metabolize fructose (fructokinase knockout mice). Of interest, our data indicate that despite their inability to taste sweetness, P2X2/3 DKO mice still prefer caloric sugars (including fructose and glucose) to water in long-term testing, although with diminished preference compared with control mice. Despite reduced intake of caloric sugars by P2X2/3 DKO animals, the DKO mice still show increased levels of the sugar-dependent hormone FGF21 (fibroblast growth factor 21) in plasma and liver. Despite lower sugar intake, taste-blind mice develop severe features of metabolic syndrome due to reduced sensitivity to leptin, reduced ability to mobilize and oxidize fats, and increased hepatic de novo lipogenesis. In contrast to P2X2/3 DKO and wild-type mice, fructokinase knockout mice, which cannot metabolize fructose and are protected against fructose-induced metabolic syndrome, demonstrate reduced preference and intake for all fructose-containing sugars tested but not for glucose or artificial sweeteners. Based on these observations, we conclude that sugar can induce metabolic syndrome in mice independently of its sweet properties. Furthermore, our data demonstrate that the metabolism of fructose is necessary for sugar to drive intake and preference in mice.
Sigma-Aldrich
Anti-KHK antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
Supelco
Diaion® HP-20, pkg of 1000 g