MilliporeSigma
  • Synthetic protection short interfering RNA screen reveals glyburide as a novel radioprotector.
Synthetic protection short interfering RNA screen reveals glyburide as a novel radioprotector.
Radiation research (2009-09-24)
Jianfei Jiang, Peter R McDonald, Tracy M Dixon, Darcy Franicola, Xichen Zhang, Suhua Nie, Laura D Epperly, Zhentai Huang, Valerian E Kagan, John S Lazo, Michael W Epperly, Joel S Greenberger
ABSTRACT
To assist in screening existing drugs for use as potential radioprotectors, we used a human unbiased 16,560 short interfering RNA (siRNA) library targeting the druggable genome. We performed a synthetic protection screen that was designed to identify genes that, when silenced, protected human glioblastoma T98G cells from gamma-radiation-induced cell death. We identified 116 candidate protective genes, then identified 10 small molecule inhibitors of 13 of these candidate gene products and tested their radioprotective effects. Glyburide, a clinically used second-generation hypoglycemic drug, effectively decreased radiation-induced cell death in several cell lines including T98G, glioblastoma U-87 MG, and normal lung epithelial BEAS-2B and in primary cultures of astrocytes. Glyburide significantly increased the survival of 32D cl3 murine hematopoietic progenitor cells when administrated before irradiation. Glyburide was radioprotective in vivo (90% of C57BL/6NHsd female mice pretreated with 10 mg/kg glyburide survived 9.5 Gy total-body irradiation compared to 42% of irradiated controls, P = 0.0249). These results demonstrate the power of unbiased siRNA synthetic protection screening with a druggable genome library to identify new radioprotectors.
Sigma-Aldrich
Anti-Actin antibody, Mouse monoclonal, clone AC-40, purified from hybridoma cell culture