MilliporeSigma
  • Development and Application of TK6-derived Cells Expressing Human Cytochrome P450s for Genotoxicity Testing.
Development and Application of TK6-derived Cells Expressing Human Cytochrome P450s for Genotoxicity Testing.
Toxicological sciences : an official journal of the Society of Toxicology (2020-03-12)
Xilin Li, Si Chen, Xiaoqing Guo, Qiangen Wu, Ji-Eun Seo, Lei Guo, Mugimane G Manjanatha, Tong Zhou, Kristine L Witt, Nan Mei
ABSTRACT
Metabolism plays a key role in chemical genotoxicity; however, most mammalian cells used for in vitro genotoxicity testing lack effective metabolizing enzymes. We recently developed a battery of TK6-derived cell lines that individually overexpress 1 of 8 cytochrome P450s (CYP1A1, 1A2, 1B1, 2A6, 2B6, 2C9, 2C19, and 3A4) using a lentiviral expression system. The increased expression and metabolic function of each individual CYP in each established cell line were confirmed using real-time PCR, Western blotting, and mass spectrometry analysis; the parental TK6 cells and empty vector (EV) transduced cells had negligible CYP levels. Subsequently, we evaluated these cell lines using 2 prototypical polyaromatic hydrocarbon mutagens, 7,12-dimethylbenz[a]anthracene (DMBA) and benzo[a]pyrene (B[a]P), that require metabolic activation to exert their genotoxicity. DMBA-induced cytotoxicity, phosphorylation of histone H2A.X, and micronucleus formation were significantly increased in TK6 cells with CYP1A1, 1B1, 2B6, and 2C19 expression as compared with EV controls. B[a]P significantly increased cytotoxicity, DNA damage, and chromosomal damage in TK6 cells overexpressing CYP1A1 and 1B1 when compared with EV controls. B[a]P also induced micronucleus formation in TK6 cells expressing CYP1A2. These results suggest that our CYP-expressing TK6 cell system can be used to detect the genotoxicity of compounds requiring metabolic transformation.
Sigma-Aldrich
7-Ethoxy-4-(trifluoromethyl)coumarin, ≥98% (TLC)