MilliporeSigma
  • Synthesis of peptide nucleic acid FRET probes via an orthogonally protected building block for post-synthetic labeling of peptide nucleic acids at the 5-position of uracil.
Synthesis of peptide nucleic acid FRET probes via an orthogonally protected building block for post-synthetic labeling of peptide nucleic acids at the 5-position of uracil.
Bioconjugate chemistry (2008-10-04)
Bereket Y Oquare, John-Stephen Taylor
ABSTRACT
We report the design and synthesis of an orthogonally protected peptide nucleic acid (PNA) building block, Fmoc-PNA-U'-(Dde)-OH, and its use in the construction of PNA FRET probes. This building block allows for the post-synthetic attachment of reporter groups to the amino group attached to the 5-position of uracil (U) following selective deprotection of the Dde group. We illustrate the use of this building block for the synthesis of a series of FAM Cy5 donor acceptor pairs and their ability to detect a target DNA sequence.
Sigma-Aldrich
Hydroxylamine hydrochloride, 99.999% trace metals basis
Sigma-Aldrich
Hydroxylamine hydrochloride, 99.995% trace metals basis
Sigma-Aldrich
Hydroxylamine hydrochloride, puriss. p.a., ACS reagent, ≥99.0% (RT)
Sigma-Aldrich
Hydroxylamine hydrochloride, ACS reagent, 98.0%
Sigma-Aldrich
Hydroxylamine hydrochloride, ReagentPlus®, 99%