MilliporeSigma
  • Assay of phosphotyrosyl protein phosphatase using synthetic peptide 1142-1153 of the insulin receptor.
Assay of phosphotyrosyl protein phosphatase using synthetic peptide 1142-1153 of the insulin receptor.
FEBS letters (1988-09-12)
M J King, G J Sale
ABSTRACT
Synthetic peptide 1142-1153 of the insulin receptor was phosphorylated on tyrosine by the insulin receptor and found to be a potent substrate for dephosphorylation by rat liver particulate and soluble phosphotyrosyl protein phosphatases. Apparent Km values were approximately 5 microM. Vm values (nmol phosphate removed/min per mg protein) were 0.62 (particulate) and 0.2 (soluble). This corresponds to 80% of total activity being membrane-associated, indicating that membrane-bound phosphatases are important receptor phosphatases. The phosphatase activities were distinct from acid and alkaline phosphatase. In conclusion peptide 1142-1153 provides a useful tool for the further study and characterization of phosphotyrosyl protein phosphatases.