MilliporeSigma
The Forgotten Pyrazines: Exploring the Dakin-West Reaction.
Chemistry (Weinheim an der Bergstrasse, Germany) (2020-03-28)
Martien A Würdemann, Cristina Niţu, Stefaan M A De Wildeman, Katrien V Bernaerts, Romano V A Orru
ABSTRACT
Pyrazines are an underreported class of N-heterocycles available from nitrogen-rich biomass presenting an interesting functional alternative for current aromatics. In this work, access to pyrazines obtained from amino acids by using the 90 year old Dakin-West reaction was explored. After a qualitative screening several functional proteinogenic amino acids proved good substrates for this reaction, which were successfully scaled to multigram scale synthesis of the corresponding intermediate α-acetamido ketones. Subsequently, the conditions towards pyrazine formation using δ-amino-levulinic acid were optimized, and these were employed to synthesize a relevant set of five functional dimethylpyrazines in high purity. These pyrazines can be considered a versatile toolbox of aromatic building blocks for a wide range of applications, such as in the synthesis of polymers or metal-organic frameworks.
Sigma-Aldrich
2-(Dimethylamino)pyridine, 97%
Sigma-Aldrich
DL-Cysteine, technical grade