MilliporeSigma
  • Characterization of Resveratrol, Oxyresveratrol, Piceatannol and Roflumilast as Modulators of Phosphodiesterase Activity. Study of Yeast Lifespan.
Characterization of Resveratrol, Oxyresveratrol, Piceatannol and Roflumilast as Modulators of Phosphodiesterase Activity. Study of Yeast Lifespan.
Pharmaceuticals (Basel, Switzerland) (2020-09-03)
Adrián Matencio, Francisco García-Carmona, José Manuel López-Nicolás
ABSTRACT
Our desire to live longer has led to an ever-increasing number of novel antiaging products. However, few molecules have any real effect and new ones need to be studied before they can be used commercially. In this contribution, activation of the caloric restriction (CR) pathway was studied using different three (resveratrol, oxyresveratrol and piceatannol)-a family with demonstrated bioactivity on phosphodiesterase activity. The high-affinity phosphodiesterase type 2 (PDE2) of Saccharomyces cerevisiae was expressed in Escherichia coli, purified and characterized. The activity and the inhibitory activity of each stilbene was studied, and the findings were compared in vitro and in silico with those obtained with roflumilast-a human PDE4 inhibitor widely used in chronic obstructive pulmonary diseases. Finally, an in vivo chronological lifespan assay using WT S. cerevisiae and ΔPDE2 S. cerevisiae strains was carried out. It was demonstrated that stilbenes can modulate yPDE2 activity, increasing the lifespan of the yeast by 18% over a control (in combination with other pathways). In addition, roflumilast increased the lifespan in the WT strain. The findings as a whole would increase the range of lifespan products available and suggest novel uses for approved drugs.
Sigma-Aldrich
Piceatannol, powder
Sigma-Aldrich
Yeast Nitrogen Base Without Amino Acids, Yeast classification medium used for selecting yeasts based on amino acid and carbohydrate requirements
Sigma-Aldrich
Benzamidine, ≥95.0%
Sigma-Aldrich
L-(−)-Glucose, ≥99%
Sigma-Aldrich
Adenosine 3′,5′-cyclic monophosphate tris salt, ≥97% (HPLC), powder
Sigma-Aldrich
Oxyresveratrol, ≥97.0% (HPLC)