MilliporeSigma
  • Self-Assembled Quaternary Ammonium-Containing Comb-Like Polyelectrolytes for the Hydrolysis of Organophosphorous Esters: Effect of Head Groups and Counter-Ions.
Self-Assembled Quaternary Ammonium-Containing Comb-Like Polyelectrolytes for the Hydrolysis of Organophosphorous Esters: Effect of Head Groups and Counter-Ions.
ChemPlusChem (2020-09-01)
Tatiana N Pashirova, Petr A Fetin, Alexey A Lezov, Matvey V Kadnikov, Farida G Valeeva, Evgenia A Burilova, Alexander Yu Bilibin, Ivan M Zorin
ABSTRACT
The aim of this work was to increase the efficiency of catalytic systems for the hydrolytic cleavage of 4-nitrophenyl esters of phosphonic acids. Quaternary ammonium-containing comb-like polyelectrolytes («polymerized micelles») with ester cleavable fragments and a low aggregation threshold were used as catalysts. The synthesis of poly(11-acryloyloxyundecylammonium) surfactants with different counterions (Br- , NO3- , CH3 C6 H4 SO3- ) and head groups was realized by micellar free-radical polymerization. Molecular weight, critical association concentration, particle sizes and solubilization properties toward Orange OT were determined. Self-assemblies organized by poly(11-acryloyloxyundecyltrimethyl ammonium) bromide successfully catalyze the hydrolysis of 4-nitrophenyl butylchloromethylphosphonate up to two orders of magnitude compared to aqueous alkaline hydrolysis. The development of these catalysts is promising for industrial applications and organophosphorus compound detoxification.
Sigma-Aldrich
Pyrene, sublimed grade, 99%
Sigma-Aldrich
11-Bromo-1-undecanol, 98%