MilliporeSigma
  • Murrayafoline A attenuates the Wnt/beta-catenin pathway by promoting the degradation of intracellular beta-catenin proteins.
Murrayafoline A attenuates the Wnt/beta-catenin pathway by promoting the degradation of intracellular beta-catenin proteins.
Biochemical and biophysical research communications (2009-12-08)
Hyuk Choi, Jungsug Gwak, Munju Cho, Min-Jung Ryu, Jee-Hyun Lee, Sang Kyum Kim, Young Ho Kim, Gye Won Lee, Mi-Young Yun, Nguyen Manh Cuong, Jae-Gook Shin, Gyu-Yong Song, Sangtaek Oh
ABSTRACT
Molecular lesions in Wnt/beta-catenin signaling and subsequent up-regulation of beta-catenin response transcription (CRT) occur frequently during the development of colon cancer. To identify small molecules that suppress CRT, we screened natural compounds in a cell-based assay for detection of TOPFalsh reporter activity. Murrayafoline A, a carbazole alkaloid isolated from Glycosmis stenocarpa, antagonized CRT that was stimulated by Wnt3a-conditioned medium (Wnt3a-CM) or LiCl, an inhibitor of glycogen synthase kinase-3beta (GSK-3beta), and promoted the degradation of intracellular beta-catenin without altering its N-terminal phosphorylation at the Ser33/37 residues, marking it for proteasomal degradation, or the expression of Siah-1, an E3 ubiquitin ligase. Murrayafoline A repressed the expression of cyclin D1 and c-myc, which is known beta-catenin/T cell factor (TCF)-dependent genes and thus inhibited the proliferation of various colon cancer cells. These findings indicate that murrayafoline A may be a potential chemotherapeutic agent for use in the treatment of colon cancer.
Sigma-Aldrich
Murrayafoline A, ≥90% (LC/MS-ELSD)