MilliporeSigma
  • Comparison effects of olive leaf extract and oleuropein compounds on male reproductive function in cyclophosphamide exposed mice.
Comparison effects of olive leaf extract and oleuropein compounds on male reproductive function in cyclophosphamide exposed mice.
Heliyon (2020-04-28)
Ayoob Rostamzadeh, Hossein Amini-Khoei, Mohammad Javad Mardani Korani, Mohammad Rahimi-Madiseh
ABSTRACT
Spermatogenesis is a complicated process in which sperm is susceptible to various chemotherapy drugs such as cyclophosphamide (CP). As olive leaf extract (OLE) and its active ingredient, oleuropein, have variousantioxidant, anti-apoptotic, and anti-inflammatory properties the aim of the present study was to investigate the effects of OLE and oleuropein on male reproductive function focusing antioxidative effects and histological modifications in the testes of CP-exposed mice. In order to do this, 80 NMRI male mice were divided into eight groups including control group, group received CP, group received OLE, group received oleuropein, group received OLE following CP exposure, group received oleuropein following CP exposure, group received OLE plus oleuropein and group received OLE plus oleuropein following CP exposure. In all groups CP (single dose of 100 mg/kg (, OLE (100 mg/kg for consequence 28 days) and oleuropein (100 mg/kg for consequence 28 days) were injected intraperitoneally. Moreover, testis histology, sperm parameters and serum levels of LH, FSH, MDA and antioxidant capacity were investigated. Results showed that CP caused oxidative state and abnormal changes in sperms and testes. Besides, treatments with oleuropein and OLE led to mitigate the harmful effects of CP on the male reproductive system. In conclusion, our findings showed that olive's compounds can diminish the hazardous effects of CP on spermatogenesis in mice.
Sigma-Aldrich
Oleuropein, ≥98.0%