MilliporeSigma
  • Antidepressant-like effects of the novel, selective, 5-HT2C receptor agonist WAY-163909 in rodents.
Antidepressant-like effects of the novel, selective, 5-HT2C receptor agonist WAY-163909 in rodents.
Psychopharmacology (2007-02-14)
Sharon Rosenzweig-Lipson, Annmarie Sabb, Gary Stack, Paul Mitchell, Irwin Lucki, Jessica E Malberg, Steve Grauer, Julie Brennan, John F Cryan, Stacey J Sukoff Rizzo, John Dunlop, James E Barrett, Karen L Marquis
ABSTRACT
Activation of one or more of the serotonin (5-HT) receptors may play a role in mediating the antidepressant effects of SSRIs. The present studies were conducted to evaluate the effects of the novel 5-HT2C receptor agonist WAY-163909 in animal models of antidepressant activity (forced swim test (FST), resident-intruder, olfactory bulbectomy (BULB)), in a schedule-induced polydipsia (SIP) model of obsessive-compulsive disorder and in a model for evaluating sexual dysfunction. WAY-163909 (10 mg/kg, i.p. or s.c.) decreased immobility time in Wistar-Kyoto rats in the FST, effects that were reversed by the 5-HT2C/2B receptor antagonist SB 206553. Moreover, in Sprague-Dawley rats, the profile of WAY-163909 (decreased immobility, increased swimming) in the FST was comparable to the effects of SSRIs. Acute treatment with WAY-163909 (0.33 mg/kg, s.c.) decreased rodent aggression at doses lower than those required for decreasing total behavior. Administration of WAY-163909 (3 mg/kg, i.p.) for 5 or 21 days decreased the BULB-induced hyperactivity in rats. Additionally, acute administration of WAY-163909 (3 mg/kg, i.p.) decreased adjunctive drinking in a SIP model. The effects of WAY-163909 were reversed by the 5-HT(2C/2B) receptor antagonist SB 206553 and the selective 5-HT2C receptor antagonist SB 242084. Chronic administration of WAY-163909 produced deficits in sexual function at doses higher (10 mg/kg, i.p.) than those required for antidepressant-like effects in the BULB model. Taken together, these results demonstrate that the novel 5-HT2C receptor agonist WAY-163909 produces rapid onset antidepressant-like effects in animal models and may be a novel treatment for depression.
Sigma-Aldrich
WAY-163909 hydrochloride, ≥98% (HPLC)