MilliporeSigma
  • A Highly Active Ylide-Functionalized Phosphine for Palladium-Catalyzed Aminations of Aryl Chlorides.
A Highly Active Ylide-Functionalized Phosphine for Palladium-Catalyzed Aminations of Aryl Chlorides.
Angewandte Chemie (International ed. in English) (2018-11-20)
Philip Weber, Thorsten Scherpf, Ilja Rodstein, Dominik Lichte, Lennart T Scharf, Lukas J Gooßen, Viktoria H Gessner
ABSTRACT
Ylide-functionalized phosphine ligands (YPhos) were rationally designed to fit the requirements of Buchwald-Hartwig aminations at room temperature. This ligand class combines a strong electron-donating ability comparable to NHC ligands with high steric demand similar to biaryl phosphines. The active Pd species are stabilized by agostic C-H⋅⋅⋅Pd rather than by Pd-arene interactions. The practical advantage of YPhos ligands arises from their easy and scalable synthesis from widely available, inexpensive starting materials. Benchmark studies showed that YPhos-Pd complexes are superior to the best-known phosphine ligands in room-temperature aminations of aryl chlorides. The utility of the catalysts was demonstrated by the synthesis of various arylamines in high yields within short reaction times.
Sigma-Aldrich
joYPhos, Umicore
Sigma-Aldrich
keYPhos
Sigma-Aldrich
trYPhos, Umicore