MilliporeSigma
  • Disruption of mitochondrial malate-aspartate shuttle activity in mouse blastocysts impairs viability and fetal growth.
Disruption of mitochondrial malate-aspartate shuttle activity in mouse blastocysts impairs viability and fetal growth.
Biology of reproduction (2008-10-31)
Megan Mitchell, Kara S Cashman, David K Gardner, Jeremy G Thompson, Michelle Lane
ABSTRACT
The nutrient requirements and metabolic pathways used by the developing embryo transition from predominantly pyruvate during early cleavage stages to glucose at the blastocyst; however, the complexities involved in the regulation of metabolism at different developmental stages are not clear. The aims of this study were to examine the role of the malate-aspartate shuttle (MAS) in nutrient metabolism pathways in the developing mouse blastocyst and the consequences of impaired metabolism on embryo viability and fetal and placental growth. Eight-cell-stage mouse embryos were cultured in the presence of the MAS inhibitor amino-oxyacetate, with or without pyruvate as an energy substrate in the media. When the MAS was inhibited, the rate of glycolysis and lactate production was significantly elevated and glucose uptake reduced, relative to control cultured embryos in the presence of pyruvate. Despite these changes in embryo metabolism, this did not influence development to the blastocyst stage, but it did reduce the number of inner cell mass and trophectoderm cells. When these embryos were transferred to psuedopregnant females, inhibition of the MAS significantly reduced the proportion of embryos that implanted and developed into fetuses on Day 18 of pregnancy. Finally, fetal growth was reduced while placental weight was maintained, leading to a decreased fetal:placental weight ratio relative to control embryos. These results suggest that impaired metabolism of glucose in the blastocyst via the MAS alters the ability of the embryos to implant and form a pregnancy and leads to reduced fetal weight, likely via altered placental development and function.
Product Description
Supelco
Pyrene, analytical standard
Pyrene, BCR®, certified reference material
Sigma-Aldrich
Pyrene, puriss. p.a., for fluorescence, ≥99.0% (GC)
Sigma-Aldrich
Pyrene, 98%
Sigma-Aldrich
Pyrene, sublimed grade, 99%
Supelco
Pyrene, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Sigma-Aldrich
Phenol Red, ACS reagent
Sigma-Aldrich
Phenol Red, powder, BioReagent, suitable for cell culture