MilliporeSigma
  • Activation of protein kinase C modulates BACE1-mediated beta-secretase activity.
Activation of protein kinase C modulates BACE1-mediated beta-secretase activity.
Neurobiology of aging (2006-12-13)
Lizhen Wang, Hoon Shim, Chengsong Xie, Huaibin Cai
ABSTRACT
beta-Site APP cleavage enzyme 1 (BACE1) is the beta-secretase responsible for generating amyloid-beta (A beta) peptides in Alzheimer's disease (AD). Previous studies suggest that activation of protein kinase C (PKC) modulates the beta-secretase-mediated cleavage of APP and reduces the production of A beta. The mechanism of PKC-mediated modulation of beta-secretase activity, however, remains elusive. We report here that activation of PKC modulated beta-secretase activity through either suppressing the accumulation or promoting the translocation of BACE1 protein in a cell type-dependent manner. We found that activation of PKC suppressed the accumulation of BACE1 protein in fibroblasts through an enhancement of intracellular protease activities. In neurons, activation of PKC did not alter the expression level of BACE1, but led to more BACE1 translocated to the cell surface, resulting in a decreased cleavage of APP at the beta1 site. Together, Our findings provide novel mechanisms of PKC-mediated modulation of beta-secretase activity, suggesting that alteration of the intracellular trafficking of BACE1 may serve as a useful therapeutic strategy to lower the production of A beta in AD.
Product Description
SAFC
Basal Medium Eagle, With Earle′s salts and sodium bicarbonate, without L-glutamine, liquid, sterile-filtered, suitable for cell culture
Sigma-Aldrich
Basal Medium Eagle, With Earle′s salts and L-glutamine, without sodium bicarbonate, powder, suitable for cell culture
Sigma-Aldrich
Phorbol 12-myristate 13-acetate, ≥99% (TLC), film or powder
Sigma-Aldrich
Chloroquine diphosphate salt, powder or crystals, 98.5-101.0% (EP)