MilliporeSigma
  • Identification of Required Host Factors for SARS-CoV-2 Infection in Human Cells.
Identification of Required Host Factors for SARS-CoV-2 Infection in Human Cells.
Cell (2020-11-05)
Zharko Daniloski, Tristan X Jordan, Hans-Hermann Wessels, Daisy A Hoagland, Silva Kasela, Mateusz Legut, Silas Maniatis, Eleni P Mimitou, Lu Lu, Evan Geller, Oded Danziger, Brad R Rosenberg, Hemali Phatnani, Peter Smibert, Tuuli Lappalainen, Benjamin R tenOever, Neville E Sanjana
ABSTRACT
To better understand host-virus genetic dependencies and find potential therapeutic targets for COVID-19, we performed a genome-scale CRISPR loss-of-function screen to identify host factors required for SARS-CoV-2 viral infection of human alveolar epithelial cells. Top-ranked genes cluster into distinct pathways, including the vacuolar ATPase proton pump, Retromer, and Commander complexes. We validate these gene targets using several orthogonal methods such as CRISPR knockout, RNA interference knockdown, and small-molecule inhibitors. Using single-cell RNA-sequencing, we identify shared transcriptional changes in cholesterol biosynthesis upon loss of top-ranked genes. In addition, given the key role of the ACE2 receptor in the early stages of viral entry, we show that loss of RAB7A reduces viral entry by sequestering the ACE2 receptor inside cells. Overall, this work provides a genome-scale, quantitative resource of the impact of the loss of each host gene on fitness/response to viral infection.
Sigma-Aldrich
Amlodipine besylate, ≥98% (HPLC)
Roche
KAPA SYBR® FAST One-Step, suitable for RT-PCR, 2 ×, Universal
Sigma-Aldrich
Anti-2A peptide Antibody, clone 3H4, clone 3H4, from mouse