MilliporeSigma
  • A 12p13 GRIN2B deletion is associated with developmental delay and macrocephaly.
A 12p13 GRIN2B deletion is associated with developmental delay and macrocephaly.
Human genome variation (2016-09-23)
Naoya Morisada, Tomoaki Ioroi, Mariko Taniguchi-Ikeda, Ming Juan Ye, Nobuhiko Okamoto, Toshiyuki Yamamoto, Kazumoto Iijima
ABSTRACT
N-methyl D-aspartate receptor subtype 2B (GluN2B), encoded by GRIN2B, is one of the components of the N-methyl D-aspartate receptor protein. Aberrations in GRIN2B have been reported to be responsible for various types of neurodevelopmental disorders. We report a Japanese boy with an ~2 Mb interstitial deletion in 12p13 involving the entire GRIN2B gene, who presented with intellectual disability, motor developmental delay and marked macrocephaly.