MilliporeSigma
  • Efficacy of the PSD95 inhibitor Tat-NR2B9c in mice requires dose translation between species.
Efficacy of the PSD95 inhibitor Tat-NR2B9c in mice requires dose translation between species.
Journal of cerebral blood flow and metabolism : official journal of the International Society of Cerebral Blood Flow and Metabolism (2015-12-15)
Lucy M Teves, Hong Cui, Michael Tymianski
ABSTRACT
Tat-NR2B9c, a clinical-stage stroke neuroprotectant validated in rats and primates, was recently deemed ineffective in mice. To evaluate this discrepancy, we conducted studies in mice subjected to temporary middle cerebral artery occlusion (tMCAO) for either 30 or 60 min according to the established principles for dose-translation between species. Tat-NR2B9c treatment reduced infarct volume by by 24.5% (p = 0.49) and 26.0% (p = 0.03) for 30 and 60 min tMCAO, respectively, at the rat-equivalent dose of 10 nMole/g, but not at the previously reported 3 nMole/g in mice. Dose translation is thus critical when preclinical experiments are conducted in new species.
Sigma-Aldrich
Tat-NR2B9c trifluoroacetate, ≥95% (HPLC)