MilliporeSigma
Identification of Hsp90 Inhibitors with Anti-Plasmodium Activity.
Antimicrobial agents and chemotherapy (2018-01-18)
Dora Posfai, Amber L Eubanks, Allison I Keim, Kuan-Yi Lu, Grace Z Wang, Philip F Hughes, Nobutaka Kato, Timothy A Haystead, Emily R Derbyshire
ABSTRACT
Malaria remains a global health burden partly due to Plasmodium parasite resistance to first-line therapeutics. The molecular chaperone heat shock protein 90 (Hsp90) has emerged as an essential protein for blood-stage Plasmodium parasites, but details about its function during malaria's elusive liver stage are unclear. We used target-based screens to identify compounds that bind to Plasmodium falciparum and human Hsp90, which revealed insights into chemotypes with species-selective binding. Using cell-based malaria assays, we demonstrate that all identified Hsp90-binding compounds are liver- and blood-stage Plasmodium inhibitors. Additionally, the Hsp90 inhibitor SNX-0723 in combination with the phosphatidylinositol 3-kinase inhibitor PIK-75 synergistically reduces the liver-stage parasite load. Time course inhibition studies with the Hsp90 inhibitors and expression analysis support a role for Plasmodium Hsp90 in late-liver-stage parasite development. Our results suggest that Plasmodium Hsp90 is essential to liver- and blood-stage parasite infections and highlight an attractive route for development of species-selective PfHsp90 inhibitors that may act synergistically in combination therapies to prevent and treat malaria.
Sigma-Aldrich
Geldanamycin-FITC, ≥98% (HPLC), from Streptomyces hygroscopicus