MilliporeSigma
  • Domain structure and dynamics in the helical filaments formed by RecA and Rad51 on DNA.
Domain structure and dynamics in the helical filaments formed by RecA and Rad51 on DNA.
Proceedings of the National Academy of Sciences of the United States of America (2001-07-19)
X Yu, S A Jacobs, S C West, T Ogawa, E H Egelman
ABSTRACT
Both the bacterial RecA protein and the eukaryotic Rad51 protein form helical nucleoprotein filaments on DNA that catalyze strand transfer between two homologous DNA molecules. However, only the ATP-binding cores of these proteins have been conserved, and this same core is also found within helicases and the F1-ATPase. The C-terminal domain of the RecA protein forms lobes within the helical RecA filament. However, the Rad51 proteins do not have the C-terminal domain found in RecA, but have an N-terminal extension that is absent in the RecA protein. Both the RecA C-terminal domain and the Rad51 N-terminal domain bind DNA. We have used electron microscopy to show that the lobes of the yeast and human Rad51 filaments appear to be formed by N-terminal domains. These lobes are conformationally flexible in both RecA and Rad51. Within RecA filaments, the change between the "active" and "inactive" states appears to mainly involve a large movement of the C-terminal lobe. The N-terminal domain of Rad51 and the C-terminal domain of RecA may have arisen from convergent evolution to play similar roles in the filaments.
Sigma-Aldrich
RAD51 human, recombinant, expressed in E. coli, ≥80% (SDS-PAGE)