MilliporeSigma
  • Antifungal Halogenated Cyclopentenones from the Endophytic Fungus Saccharicola bicolor of Bergenia purpurascens by the One Strain-Many Compounds Strategy.
Antifungal Halogenated Cyclopentenones from the Endophytic Fungus Saccharicola bicolor of Bergenia purpurascens by the One Strain-Many Compounds Strategy.
Journal of agricultural and food chemistry (2019-12-10)
Min Zhao, Da-Le Guo, Guang-Hui Liu, Xian Fu, Yu-Cheng Gu, Li-Sheng Ding, Yan Zhou
ABSTRACT
Phytopathogenic fungi have been considered as being an enormous threat in the agricultural system. In our search of new antifungal natural products, nine new halogenated cyclopentenones, bicolorins A-I (1-3, and 5-10), along with three known cyclopentenones (4, 11, and 12) were isolated from the endophytic fungus Saccharicola bicolor of Bergenia purpurascens by the one strain-many compounds strategy. Their structures and absolute configurations were elucidated based on extensive spectroscopic analysis, X-ray crystallographic analysis, and time-dependent density functional theory-equivalent circulating density calculations. Compounds 1-12 showed antifungal activities against five phytopathogenic fungi (Uromyces viciae-fabae, Pythium dissimile, Gibberella zeae, Aspergillus niger, and Sclerotinia sclerotiorum). Especially, bicolorins B and D (2 and 5) exhibited strong antifungal activities against P. dissimile with the MIC values of 6.2 and 8.5 μg/mL, respectively, compared with the positive control cycloheximide (MIC of 8.6 μg/mL). Additionally, bicolorin D was proven to be potently antifungal against S. sclerotiorum in vitro and in vivo. This work provides an effective strategy for searching antifungal candidate agents.